EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Effects of hydrophobic and ionic interactions on glycation of casein during Maillard reaction.
PMID 25363663
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32900
Benalaxyl-M, PESTANAL®, analytical standard
C20H23NO3