EMAIL THIS PAGE TO A FRIEND
The Journal of invasive cardiology
Outcome of patients on oral anticoagulation undergoing coronary artery stenting: data from discharge to 12 months in the Warfarin and Coronary Stenting (WAR-STENT) Registry.
PMID 25363997
Abstract
To obtain further, and more focused, information on the efficacy and safety of the antithrombotic regimens, including triple therapy (TT) of warfarin, aspirin, and clopidogrel; dual therapy (DT) of warfarin and single antiplatelet agent (aspirin or clopidogrel); and dual-antiplatelet therapy (DAPT) of aspirin and clopidogrel, prescribed to patients on oral anticoagulation (OAC) undergoing percutaneous coronary intervention with stent (PCI-S). The true efficacy and safety of TT, DT, and DAPT in OAC patients undergoing PCI-S is largely undefined. We analyzed the database of the prospective, multicenter WARfarin and coronary STENTing (WAR-STENT) registry (ClinicalTrials.gov identifier NCT00722319), only including the post-discharge period. Of the 401 patients discharged alive from index hospitalization, 339 (85%), 20 (5%), and 42 (10%) were prescribed TT, DT, and DAPT, respectively. Throughout a mean follow-up of 368.3 ± 22.8 days, major adverse cardiovascular events (MACE) (including cardiovascular death, myocardial infarction, repeat revascularization, stent thrombosis, and thromboembolism), total bleeding, major bleeding, and combination of MACE plus total bleeding were comparable across the three treatment groups. The absolute rate of major bleeding with TT was 4%. The antithrombotic treatment actually ongoing at major bleeding was TT in 44%, DT in 50%, and DAPT in 6% of cases. In the real-world population of OAC patients undergoing PCI-S in the WAR-STENT registry, the three antithrombotic regimens of TT, DT, and DAPT showed comparable efficacy and safety. Due to several limitations, our data cannot be considered conclusive in confuting the current recommendations to prescribe TT. Further properly designed and sized studies are warranted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
30640
1-Decanethiol, technical, ≥95% (GC)
C10H22S