EMAIL THIS PAGE TO A FRIEND
Growth factors (Chur, Switzerland)
Effects of anti-aggregant, anti-inflammatory and anti-coagulant drug consumption on the preparation and therapeutic potential of plasma rich in growth factors (PRGF).
PMID 25365465
Abstract
The prevalence and incidence of trauma-related injuries, coronary heart disease and other chronic diseases increase dramatically with age. This population sector is therefore a regular consumer of different types of drugs that may affect platelet aggregation and the coagulation cascade. We have evaluated whether the consumption of acetylsalicylic acid, acenocoumarol, glucosamine sulfate and chondroitin sulfate, and therefore their presence in blood, could interfere with the preparation and biological outcomes of plasma rich in growth factors (PRGF). Clotting time, clot retraction and platelet activation of PRGF was evaluated. PRGF growth factor content and the release of different biomolecules by tendon fibroblasts were also quantified, as well as cell proliferation and cell migration. The preparation and biological potential of PRGF is not affected by the intake of the evaluated drugs, and solely its angiogenic potential and its capacity to induce HA and fibronectin synthesis, is reduced in patients taking anti-coagulants.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01905
Adenosine 5′-diphosphate, ≥95% (HPLC)
C10H15N5O10P2