EMAIL THIS PAGE TO A FRIEND
International urogynecology journal
Differences in recurrent prolapse at 1 year after total vs supracervical hysterectomy and robotic sacrocolpopexy.
PMID 25366305
Abstract
Optimal management of the cervix at the time of hysterectomy and sacrocolpopexy for primary uterovaginal prolapse is unknown. Our hypothesis was that recurrent prolapse at 1 year would be more likely after a supracervical robotic hysterectomy (SRH) compared with a total robotic hysterectomy (TRH) at the time of robotic sacrocolpopexy (RSCP) for uterovaginal prolapse. This was a retrospective cohort analysis of 83 women who underwent hysterectomy with RSCP over a 24-month period (40 with TRH and 43 with SRH). At 1 year post-procedure, subjects completed validated questionnaires regarding pelvic floor symptoms, sexual function, and global satisfaction, and underwent a pelvic examination to identify mesh exposure and evaluate pelvic floor support. Demographics of the two groups were similar, except for a higher mean body mass index in the TRH group (31.9 TRH vs 25.8 SRH kg/m(2), p < 0.001). The rate of recurrent prolapse ≥ stage II was higher for women who underwent SRH compared with TRH (41.9 % vs 20.0 %, p = 0.03; OR 2.8, 95 % CI, 1.07-7.7). However, when this was analyzed as recurrence ≥ hymen, there was no difference between groups (12.5 % TRH vs 18.6 % SRH, p = 0.45). Likewise, there was no difference between groups when a composite measure of success was used (30 out of 40 [75 %] TRH vs 29 out of 43 [67.4 %] SRH, p = 0.45). Women who underwent an SRH were 2.8 times more likely to have a recurrent prolapse, ≥ stage II, at 1 year, compared with those who underwent a TRH, but when composite assessment scores were used there was no difference between the groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1319
Thyrotropin releasing hormone, ≥98% (HPLC), powder
C16H22N6O4