EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
Applying 'Sequential Windowed Acquisition of All Theoretical Fragment Ion Mass Spectra' (SWATH) for systematic toxicological analysis with liquid chromatography-high-resolution tandem mass spectrometry.
PMID 25366975
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31640
4,4′-Diaminodiphenylmethane, analytical standard
C13H14N2
C0100000
Caffeine, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
1085003
Caffeine, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1009
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
56396
Caffeine, certified reference material, TraceCERT®
C8H10N4O2
Y0000787
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
1086006
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1095
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
D0899
Diazepam
C16H13ClN2O
D0940000
Diazepam, European Pharmacopoeia (EP) Reference Standard
C16H13ClN2O
D9900
Diazepam solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis
C16H13ClN2O
D-907
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C16H13ClN2O
H1512
Haloperidol, powder
C21H23ClFNO2
1303002
Haloperidol, United States Pharmacopeia (USP) Reference Standard
C21H23ClFNO2
H100
Haloperidol
C21H23ClFNO2
H0100000
Haloperidol, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
Y0001518
Haloperidol for peak identification, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
Y0001517
Haloperidol for system suitability, European Pharmacopoeia (EP) Reference Standard
C21H23ClFNO2
41019
Melting point standard 235-237°C, analytical standard
C8H10N4O2
75035
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C8H10N4O2
Z103
Zolpidem, ≥98% (HPLC), solid
C19H21N3O