EMAIL THIS PAGE TO A FRIEND
Reviews of environmental contamination and toxicology
Mechanisms of hexavalent chromium resistance and removal by microorganisms.
PMID 25367133
Abstract
Chromium has been and is extensively used worldwide in multiple industrial processes and is routinely discharged to the environment from such processes. Therefore, this heavy metal is a potential threat to the environment and to public health, primarily because it is non-biodegradable and environmentally persistent. Chromium exists in several oxidation states, the most stable of which are trivalent Cr(Ill) and hexavalent Cr(VI) species. Each species possesses its own individual chemical characteristics and produces its own biological effects. For example, Cr (Ill) is an essential oligoelement for humans, whereas Cr(VI) is carcinogenic and mutagenic. Several chemical methods are used to remove Cr(VI) from contaminated sites. Each of these methods has advantages and disadvantages. Currently, bioremediation is often the preferred method to deal with Cr contaminated sites, because it is eco-friendly, cost-effective and is a "natural" technology. Many yeast, bacterial and fungal species have been assessed for their suitability to reduce or remove Cr(VI) contamination. The mechanisms by which these microorganisms resist and reduce Cr(VI) are variable and are species dependent. There are several Cr-resistance mechanisms that are displayed by microorganisms. These include active efflux of Cr compounds, metabolic reduction of Cr(VI) to Cr (ill), and either intercellular or extracellular prec1p1tation. Microbial Cr (VI) removal typically involves three stages: binding of chromium to the cell surface, translocation of chromium into the cell, and reduction of Cr(VI) to Cr (ill). Cr(VI) reduction by microorganisms may proceed on the cell surface, outside the cell, or intracellularly, either directly via chromate reductase enzymes, or indirectly via metabolite reduction of Cr(VI). The uptake of chromium ions is a biphasic process. The primary step is known as biosorption, a metabolic energyindependent process. Thereafter, bioaccumulation occurs, but is much slower, and is dependent on cell metabolic activity. Choosing an appropriate bioremediation strategy for Cr is extremely important and must involve investigating and understanding the key mechanisms that are involved in microbial resistance to and removal of Cr(VI).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27066
Chromium, coating quality, 99.95%, grit
Cr
374849
Chromium, chips, 99.995% trace metals basis
Cr
255610
Chromium, chips, thickness ~2 mm, 99.5%
Cr
266264
Chromium, powder, 99.5%, −100 mesh
Cr
266299
Chromium, powder, ≥99% trace metals basis, −325 mesh
Cr
749052
Chromium, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.95% trace metals basis
Cr
GF97385338
Chromium, foil, not light tested, 100x100mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF23770885
Chromium, foil, not light tested, 100x100mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF24605816
Chromium, foil, not light tested, 100x100mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF22618739
Chromium, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr
GF00270593
Chromium, foil, not light tested, 25x25mm, thickness 0.0015mm, permanent polyester support, 99.99+%
Cr
GF88955410
Chromium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.99+%
Cr
GF94865170
Chromium, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 99.99+%
Cr
GF73692377
Chromium, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.99+%
Cr
GF61658725
Chromium, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 99.99+%
Cr
GF81124286
Chromium, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 99.99+%
Cr
GF05241316
Chromium, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 99.99+%
Cr
GF98123250
Chromium, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 99.99+%
Cr
GF34438923
Chromium, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 99.99+%
Cr
GF49392456
Chromium, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 99.99+%
Cr
GF88985446
Chromium, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 99.99+%
Cr
GF30096053
Chromium, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF12477157
Chromium, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 99.99+%
Cr
GF84397168
Chromium, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 99.99+%
Cr
GF50354692
Chromium, foil, not light tested, 25x25mm, thickness 0.02mm, permanent polyester support, 99.99+%
Cr
GF74347050
Chromium, foil, not light tested, 25x25mm, thickness 0.03mm, permanent polyester support, 99.99+%
Cr
GF07622759
Chromium, foil, 50x50mm, thickness 3.0mm, hard, 99.95%
Cr
GF35390651
Chromium, foil, light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 99.99+%
Cr
GF46122645
Chromium, foil, not light tested, 25x25mm, thickness 0.04mm, permanent polyester support, 99.99+%
Cr
GF31792956
Chromium, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.99+%
Cr