EMAIL THIS PAGE TO A FRIEND
Journal of Translational Medicine
Specific-detection of clinical samples, systematic functional investigations, and transcriptome analysis reveals that splice variant MUC4/Y contributes to the malignant progression of pancreatic cancer by triggering malignancy-related positive feedback loops signaling.
PMID 25367394