EMAIL THIS PAGE TO A FRIEND
Medical Oncology (Northwood, London, England)
X-linked inhibitor of apoptosis-associated factor l (XAFl) enhances the sensitivity of colorectal cancer cells to cisplatin.
PMID 25367849