EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
c-Abl tyrosine kinase promotes adipocyte differentiation by targeting PPAR-gamma 2.
PMID 25368164
Abstract
Adipocyte differentiation, or adipogenesis, is a complex and highly regulated process. A recent proteomic analysis has predicted that the nonreceptor tyrosine kinase Abelson murine leukemia viral oncogene (c-Abl) is a putative key regulator of adipogenesis, but the underlying mechanism remained obscure. We found that c-Abl was activated during the early phase of mouse 3T3-L1 preadipocyte differentiation. Moreover, c-Abl activity was essential and its inhibition blocked differentiation to mature adipocytes. c-Abl directly controlled the expression and activity of the master adipogenic regulator peroxisome proliferator-activator receptor gamma 2 (PPARγ2). PPARγ2 physically associated with c-Abl and underwent phosphorylation on two tyrosine residues within its regulatory activation function 1 (AF1) domain. We demonstrated that this process positively regulates PPARγ2 stability and adipogenesis. Remarkably, c-Abl binding to PPARγ2 required the Pro12 residue that has a phenotypically well-studied common human genetic proline 12 alanine substitution (Pro12Ala) polymorphism. Our findings establish a critical role for c-Abl in adipocyte differentiation and explain the behavior of the known Pro12Ala polymorphism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB1408988
Anti-FABP4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA002188
Anti-FABP4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410359
Anti-FABP4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1410360
Anti-FABP4 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB4300636
Anti-FABP4 antibody produced in rabbit, affinity isolated antibody
SAB2101328
Anti-LCP1 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2701641
Anti-lcp1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2701743
Anti-lcp1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA019493
Anti-LCP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406091
Anti-LPL antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401231
Anti-LPL antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2700761
Anti-LPL antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2501530
Anti-PSMA4 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2104335
Anti-PSMA4, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1408986
Monoclonal Anti-FABP4 antibody produced in mouse, clone 3G1, purified immunoglobulin, buffered aqueous solution
SAB1408987
Monoclonal Anti-FABP4 antibody produced in mouse, clone 2F12-1B11, ascites fluid, solution
WH0002167M1
Monoclonal Anti-FABP4 antibody produced in mouse, clone 2H3-1G10, purified immunoglobulin, buffered aqueous solution
WH0005685M1
Monoclonal Anti-PSMA4 antibody produced in mouse, clone 2A10-E4, purified immunoglobulin, buffered aqueous solution
SAB2100743
Anti-FABP4 antibody produced in rabbit, affinity isolated antibody
SAB2101327
Anti-LCP1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB1403801
Monoclonal Anti-FABP4 antibody produced in mouse, clone 4G1, purified immunoglobulin, buffered aqueous solution
SAB2701143
Anti-LPL antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB5300056
Monoclonal Anti-LPL antibody produced in mouse, clone 2C5, ascites fluid