EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
A new approach for automated liquid-liquid extraction in a sequential injection manifold.
PMID 25370162
Abstract
A new approach for the implementation of liquid-liquid extraction in a sequential injection manifold is presented. The manifold consists of two syringe pumps and one multi-position valve. The use of a 30 % MeOH/H2O (v/v) solution as the carrier, with isobutanol as the extractant, made it possible to avoid the problems associated with the different affinities of the organic and aqueous phases for Teflon tubing, and the formation of bubbles. The suitability of the proposed method was demonstrated by the determination of thiamine in pharmaceutical preparations and dietary supplements. Detection and quantification limits were estimated as 9 and 30 μg L(-1), respectively. The repeatability was lower than 3 % and the intermediate precision (3 d) was lower than 7 %. The sample throughput was 14 samples per h. The results were in agreement with a high-performance liquid chromatography-electrospray/mass spectrometry reference method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
58448
2-Methyl-1-propanol, BioUltra, for molecular biology, ≥99.5% (GC)
C4H10O
82059
2-Methyl-1-propanol, analytical standard
C4H10O
58450
2-Methyl-1-propanol, puriss. p.a., ACS reagent, ≥99.5% (GC)
C4H10O
33064
2-Methyl-1-propanol, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
C4H10O
294829
2-Methyl-1-propanol, anhydrous, 99.5%
C4H10O
270466
2-Methyl-1-propanol, for HPLC, 99.5%
C4H10O
320048
2-Methyl-1-propanol, ACS reagent, ≥99.0%
C4H10O
538132
2-Methyl-1-propanol, 99.5%
C4H10O
A3537
5-Aminosalicylic acid, ≥99%
C7H7NO3
18858
5-Aminosalicylic acid, analytical standard
C7H7NO3
A79809
5-Aminosalicylic acid, 95%
C7H7NO3
77249
Heptafluorobutyric acid, ≥99.0% (GC)
C4HF7O2
52411
Heptafluorobutyric acid, for ion chromatography, ≥99.5% (GC)
C4HF7O2
164194
Heptafluorobutyric acid, 98%
C4HF7O2
49540
Heptafluorobutyric acid solution, 0.5 M in H2O, for ion pair chromatography
C4HF7O2
W217905
Isobutyl alcohol, ≥99%, FCC, FG
C4H10O
W217913
Isobutyl alcohol, natural, ≥99%, FCC, FG
C4H10O
PHR1060
Mesalamine, Pharmaceutical Secondary Standard; Certified Reference Material
C7H7NO3
Y0000297
Mesalazine, European Pharmacopoeia (EP) Reference Standard
C7H7NO3
Y0001574
Mesalazine for system suitability, European Pharmacopoeia (EP) Reference Standard
C7H7NO3
T4625
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
C12H17ClN4OS · HCl
T1270
Thiamine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
C12H17ClN4OS · HCl
51622
Thiamine hydrochloride, PharmaGrade, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C12H17ClN4OS · HCl
PHR1037
Thiamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C12H17ClN4OS · HCl
1656002
Thiamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C12H17ClN4OS · HCl
04573
Thiamine hydrochloride, tested according to Ph.Eur.
C12H17ClN4OS · HCl
T4562
Thiamine hydrochloride, meets USP testing specifications
C12H17ClN4OS · HCl
W332208
Thiamine hydrochloride, ≥98%, FCC, FG
C12H17ClN4OS · HCl
Y0000467
Thiamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C12H17ClN4OS · HCl
47858
Thiamine Hydrochloride (B1), analytical standard
C12H17ClN4OS · HCl