EMAIL THIS PAGE TO A FRIEND
Molecular Ecology
Ecotypes of an ecologically dominant prairie grass (Andropogon gerardii) exhibit genetic divergence across the U.S. Midwest grasslands' environmental gradient.
PMID 25370460