EMAIL THIS PAGE TO A FRIEND
Investigative Ophthalmology & Visual Science
Effects of mesenchymal stem/stromal cells on cultures of corneal epithelial progenitor cells with ethanol injury.
PMID 25370509
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219177
4,5-Dimethylthiazole, 97%
C5H7NS
W327409
4,5-Dimethylthiazole, ≥97%, FG
C5H7NS