EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
In-depth analysis of site-specific N-glycosylation in vitronectin from human plasma by tandem mass spectrometry with immunoprecipitation.
PMID 25374123