EMAIL THIS PAGE TO A FRIEND
Metabolomics : Official journal of the Metabolomic Society
Novel metabolic features in
PMID 25374488
Abstract
Expansive knowledge of bacterial metabolism has been gained from genome sequencing output, but the high proportion of genes lacking a proper functional annotation in a given genome still impedes the accurate prediction of the metabolism of a cell. To access to a more global view of the functioning of the soil bacterium