EMAIL THIS PAGE TO A FRIEND
Molecular Pharmaceutics
Synthesis of a novel cyclic prodrug of S-allyl-glutathione able to attenuate LPS-induced ROS production through the inhibition of MAPK pathways in U937 cells.
PMID 25375771