EMAIL THIS PAGE TO A FRIEND
Nature Reviews. Drug Discovery
Is there new hope for therapeutic matrix metalloproteinase inhibition?
PMID 25376097
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6627
5-Fluorouracil, ≥99% (HPLC), powder
C4H3FN2O2
03738
5-Fluorouracil, analytical standard
C4H3FN2O2
858471
5-Fluorouracil, 99%
C4H3FN2O2
11709
Barbituric acid, for spectrophotometric det. of cyanide, ≥99.5%
C4H4N2O3
185698
Barbituric acid, ReagentPlus®, 99%
C4H4N2O3
C7128
Chlorotrianisene, ~95%
C23H21ClO3
F8423
Fluorouracil, meets USP testing specifications
C4H3FN2O2
PHR1227
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
C4H3FN2O2
F0250000
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
C4H3FN2O2
1279000
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
C4H3FN2O2
746363
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Na2O4S
793531
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Na2O4S
204447
Sodium sulfate, ≥99.99% trace metals basis
Na2O4S
S5640
Sodium sulfate, ≥99.0%, plant cell culture tested
Na2O4S
S2804
Sodium sulfate, meets USP testing specifications
Na2O4S
S6547
Sodium sulfate, BioXtra, ≥99.0%
Na2O4S
71959
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Na2O4S
35896
Sodium sulfate, for pesticide residue analysis
Na2O4S
71963
Sodium sulfate, tested according to Ph.Eur., anhydrous
Na2O4S
88185
Tetrazole solution, for DNA synthesis, filtered through a 1 μm filter, ~0.45 M in acetonitrile
CH2N4