EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Influence of steviol glycosides on the stability of vitamin C and anthocyanins.
PMID 25376304
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19345
Steviol, analytical standard
C20H30O3