EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Reduction of lipid peroxidation products and advanced glycation end-product precursors by cyanobacterial aldo-keto reductase AKR3G1—a founding member of the AKR3G subfamily.
PMID 25376835
Abstract
The purpose of this study was to investigate the origin and function of the aldo-keto reductase (AKR) superfamily as enzymes involved in the detoxification of xenobiotics. We used the cyanobacterium Synechocystis sp. PCC 6803 as a model organism and sequence alignments to find bacterial AKRs with highest identity to human enzymes. Disappearance of NADPH was monitored spectrophotometrically to calculate enzymatic activity. The molecular weight of the native protein was determined by size exclusion chromatography. Substrate docking was performed by SwissDock. Sequence alignments identified the NADPH-dependent AKR3G1 having 41.5 and 40% identity with the human enzymes AKR1B1 and AKR1B10, respectively. Highest enzymatic efficiency was observed with 4-oxonon-2-enal (4-ONE; k(cat)/K(m), 561 s(-1) mM(-1)) and 4-hydroxynonenal (k(cat)/K(m), 26.5 s(-1) mM(-1)), respectively. P74308 is the most efficient enzyme for 4-ONE discovered until now. Cooperativity of this monomeric enzyme was observed with some substrates. Enzyme inactivation or oligomerization as possible explanations for nonhyperbolic enzyme kinetics were ruled out by Selwyn's test and gel filtration. The role of the little investigated carbonyl-reducing enzymes in detoxification seems to be in fact a very old process with rarely observed nonhyperbolic enzyme kinetics as an adaptation mechanism to higher concentrations of reactive oxygen species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
276286
(1R)-(−)-Camphorquinone, 99%
C10H14O2
346616
1,2-Naphthoquinone, 97%
C10H6O2
W254304
2,3-Heptanedione, ≥97%, FG
C7H12O2
75034
2,3-Hexanedione, analytical standard
C6H10O2
144169
2,3-Hexanedione, technical grade, 90%
C6H10O2
W255807
2,3-Hexanedione, ≥95%, FG
C6H10O2
W255801
2,3-Hexanedione, natural, 96%, FG
C6H10O2
75762
3-Deoxyglucosone, ≥75% (TLC)
C6H10O5
72800
4-Nitrobenzaldehyde, for spectrophotometric det. of amino sugars, ≥99.0%
C7H5NO3
130176
4-Nitrobenzaldehyde, 98%
C7H5NO3
P62402
4-Pyridinecarboxaldehyde, 97%
C6H5NO
W501808
5-(Hydroxymethyl)furfural, ≥99%, FG
C6H6O3
53407
5-(Hydroxymethyl)furfural, analytical standard
C6H6O3
H40807
5-Hydroxymethyl-2-furaldehyde, 99%
C6H6O3
94194
Butyraldehyde, analytical standard, contains ~0.1% 2,6-di-tert-butyl-4-methylphenol and ~1% water as stabilizer
C4H8O
418102
Butyraldehyde, purified by redistillation, ≥99.5%
C4H8O
538191
Butyraldehyde, ≥99.0%, dry
C4H8O
W221902
Butyraldehyde, ≥96.0%
C4H8O
W221910
Butyraldehyde, natural, FG
C4H8O
C1613 Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered
C17H18N2O6S
W228613
Cinnamaldehyde, natural, ≥95%, FG
C9H8O
72609
Coumarin, certified reference material, TraceCERT®
C9H6O2
C4261
Coumarin, ≥99% (HPLC)
C9H6O2
W526509
Coumarin, ≥98%
C9H6O2
01260595
Coumarin, primary reference standard
C9H6O2
Y0000438
Coumarin, European Pharmacopoeia (EP) Reference Standard
C9H6O2
27980
Crotonaldehyde, mixture of cis and trans, ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)
C4H6O
262668
Crotonaldehyde, predominantly trans, ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer
C4H6O
W248908
Furfural, ≥98%, FCC, FG
C5H4O2
319910
Furfural, ACS reagent, 99%
C5H4O2