EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Cell Physiology
CaV3.2 T-type Ca?? channels in H?S-mediated hypoxic response of the carotid body.
PMID 25377087
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0888
Fura 2-AM, ≥95% (HPLC)
C44H47N3O24
47989
Fura 2-AM, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
C44H47N3O24