EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Characterization and profiling of phenolic amides from Cortex Lycii by ultra-high performance liquid chromatography coupled with LTQ-Orbitrap mass spectrometry.
PMID 25377778
Abstract
Cortex Lycii, the root bark of Lycium chinense Mill. or Lycium barbarum L., is a frequently used traditional Chinese medicine. Phytochemical studies have shown that phenolic amides are not only characteristic compounds but also abundant ones in this plant. In the present study, an effective method was developed for structural characterization of phenolic amides from Cortex Lycii by ultra-high performance liquid chromatography coupled with linear ion trap Orbitrap tandem mass spectrometry. The fragmentation of 14 compounds including six cinnamic acid amides, six neolignanamides, and two lignanamides were studied systematically for the first time. It was found that, in the positive ion mode, neutral loss of the tyramide moiety (137 Da) or N-(4-aminobutyl)acetamide moiety (130 Da) were characteristic for these compounds. At least 54 phenolic amides were detected in the extract and 48 of them were characterized, among which 14 known compounds were identified unambiguously by comparing the retention time and mass spectra with those of reference compounds, and 34 components were tentatively identified based on the fragmentation patterns, exact mass, UV spectra, as well as retention time. Fifteen compounds were characterized as potential new ones. Additionally, the developed method was applied to analyze eight batches of samples collected from the northwest of China, and it was found that cinnamic acid amides were the main type of phenolic amides in Cortex Lycii. In conclusion, the identification of these chemicals provided essential data for further phytochemical studies, metabolites identification, and the quality control of Cortex Lycii.