EMAIL THIS PAGE TO A FRIEND
Neuropathology : Official Journal of the Japanese Society of Neuropathology
Frequencies of myohistological mitochondrial changes in patients with mitochondrial DNA deletions and the common m.3243A>G point mutation.
PMID 25378026