EMAIL THIS PAGE TO A FRIEND
Molecular Medicine Reports
Roscovitine-treated HeLa cells finalize autophagy later than apoptosis by downregulating Bcl?2.
PMID 25378060
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
318426
3,3′-Dihexyloxacarbocyanine iodide, 98%
C29H37IN2O2