EMAIL THIS PAGE TO A FRIEND
Molecular Cancer Therapeutics
Bicyclic peptides conjugated to an albumin-binding tag diffuse efficiently into solid tumors.
PMID 25381263
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90968
1,2-Ethanedithiol, analytical standard
C2H6S2
02390
1,2-Ethanedithiol, ≥98.0% (GC)
C2H6S2
398020
1,2-Ethanedithiol, technical grade, ≥90%
C2H6S2
657336
1,3,5-Tris(bromomethyl)benzene, 97%
C9H9Br3
D27802
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
C6H12N2
712256
5-Azidopentanoic acid, ≥97.0%
C5H9N3O2
290734
Dabco® 33-LV
C6H12N2
12804
HBTU, ≥98.0% (T)
C11H16F6N5OP
451045
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
I2
229695
Iodine, 99.999% trace metals basis
I2
326143
Iodine, ≥99.99% trace metals basis
I2
57660
Iodine, purum p.a., ≥99.5% (RT)
I2
W387304
Methyl phenyl sulfide, ≥99%
C7H8S
387649
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
C8H19N
496219
N,N-Diisopropylethylamine, 99.5%, biotech. grade
C8H19N
D125806
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
C8H19N
03440
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
C8H19N
03439
N-Ethyldiisopropylamine solution, for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
C8H19N
411027
Piperidine, ≥99.5%, purified by redistillation
C5H11N
571261
Piperidine, biotech. grade, ≥99.5%
C5H11N
80640
Piperidine, puriss. p.a., ≥99.0% (GC/T)
C5H11N
104094
Piperidine, ReagentPlus®, 99%
C5H11N
76046
Piperidine, analytical standard
C5H11N
80642
Piperidine, purum, ≥96.0% (GC/T)
C5H11N
W290807
Piperidine, ≥99%, FCC
C5H11N
80645
Piperidine solution, for peptide synthesis, 20% in DMF
C5H11N
88470
Thioanisole, ReagentPlus®, ≥99%
C7H8S
T28002
Thioanisole, ReagentPlus®, ≥99%
C7H8S
92358
Thioanisole, analytical standard
C7H8S