EMAIL THIS PAGE TO A FRIEND
Immunologic Research
Silenced suppressor of cytokine signaling 1 (SOCS1) enhances the maturation and antifungal immunity of dendritic cells in response to Candida albicans in vitro.
PMID 25381480