EMAIL THIS PAGE TO A FRIEND
Environmental monitoring and assessment
Determination of cadmium in biodiesel using microemulsion and electrothermal atomization atomic absorption spectrometry.
PMID 25381584
Abstract
This work aimed to prepare biodiesel microemulsions for the subsequent quantification of cadmium via graphite furnace atomic absorption spectrometry (GFAAS). The biodiesel samples were prepared using n-propanol as an emulsifier, 10% (v/v) nitric acid as the aqueous phase, and biodiesel. Pseudoternary phase diagrams were constructed to determine the microemulsion region with the specified components. The optimized conditions for microemulsion formation were 57.6% (v/v) n-propanol, 21.2% (v/v) biodiesel, and 21.2% (v/v) nitric acid solution. The stability of the microemulsified system was investigated using aqueous and organic standards, and the system was found to be stable for at least 240 min. The applied pyrolysis and atomization temperatures were 800 and 2000 °C, respectively, and 5 μg of aluminum was used as the chemical modifier. The obtained limits of detection and quantification were 0.2 and 0.5 μg kg(-1), respectively, and the characteristic mass was 1.6 pg. The precision, expressed as the relative standard deviation (% R.S.D., n = 10), was 2.5% for a sample with a cadmium concentration of 6.5 μg kg(-1). The accuracy was determined from addition and recovery experiments, with results varying from 93 to 108% recovery. This study demonstrates that the proposed method based on the use of a microemulsion formation in sample preparation can be applied as an efficient alternative for the determination of cadmium in biodiesel by GFAAS. Cadmium determination in biodiesel samples of different origins (soybean, corn, cotton, and sunflower) was evaluated after acid digestion using the inductively coupled plasma-mass spectrometry (ICP-MS) technique, and the obtained results were compared to the results obtained using the proposed method. The paired t test (95% confidence level) did not show significant differences. The concentrations of cadmium found ranged from 5.3 to 8.0 μg kg(-1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
265330
Cadmium, granular, ≥99%, 5-20 mesh
Cd
414891
Cadmium, granular, 30-80 mesh, ≥99%
Cd
265365
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Cd
265454
Cadmium, rod, diam. 4.0 mm, 99.999% trace metals basis
Cd
202886
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cd
348600
Cadmium, wire, diam. 1.0 mm, 99.999% trace metals basis
Cd
GF61936814
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF17721637
Cadmium, foil, 100x100mm, thickness 0.5mm, as rolled, 99.95%
Cd
GF64286602
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF30096345
Cadmium, foil, 100x100mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF10741889
Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF37649100
Cadmium, foil, 100x200mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF38583287
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cd
GF50990597
Cadmium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF72757503
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF66037754
Cadmium, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Cd
GF76768914
Cadmium, foil, 150x150mm, thickness 3.0mm, as rolled, 99.9%
Cd
GF33512688
Cadmium, foil, 200x200mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF94010453
Cadmium, foil, 200x200mm, thickness 1.0mm, as rolled, 99.99%
Cd
GF90633652
Cadmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Cd
GF06836238
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cd
GF71408763
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Cd
GF59435062
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cd
GF78263411
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cd
GF84554613
Cadmium, foil, 300x1000mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF19008817
Cadmium, foil, 300x300mm, thickness 0.25mm, as rolled, 99.95%
Cd
GF20958947
Cadmium, foil, 300x300mm, thickness 0.5mm, as rolled, 99.99%
Cd
GF15879954
Cadmium, foil, 300x300mm, thickness 2.0mm, as rolled, 99.99%
Cd
GF72851182
Cadmium, foil, 30x100mm, thickness 0.5mm, as rolled, 99.999%
Cd
GF61336997
Cadmium, wire reel, 5m, diameter 0.5mm, hard, 99.99+%
Cd