PloS one
Increased expression of cathepsin L: a novel independent prognostic marker of worse outcome in hepatocellular carcinoma patients.
PMID 25384089
Abstract
To investigate the expression and role of Cathepsin L (CTSL) in Hepatocellular carcinoma (HCC) tissue and cell line (MHCC-97H), and to evaluate the clinical and prognostic significance of CTSL protein in patients with HCC. The expression of CTSL was examined in HCC tissue and MHCC-97H cells by Western-blotting, Real-time PCR and immunohistochemical staining. Cell growth curve assay and colony formation assay were used to verify the effect of CTSL on the proliferation and tumor progression ability of MHCC-97H cells. Tumor formation assay in nude mice was used to analyze the effect of CTSL on the tumorigenicity of MHCC-97H cells. The status of CTSL protein in carcinoma tissues is much higher than that in paracarcinoma tissues. The overall survival of the patients with high CTSL expression was significantly shorter than the low CTSL expression group. high CTSL expression was significantly correlated with advanced clinical staging, histological grade and tumor recurrence. In vitro experiments demonstrated that over-expression of CTSL in MHCC-97H cells promoted cell proliferation and tumor progression ability. Down-regulation of CTSL showed the opposite effects. Over-expression of CTSL increase the tumorigenicity of MHCC-97H cells by in vivo experiments. Moreover, multivariate analysis suggested that CTSL expression might be an independent prognostic indicator for the survival of HCC patients after curative surgery. CTSL might involve in the development and progression of HCC as a oncogene, and thereby may be a valuable prognostic marker for HCC patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU106791 MISSION® esiRNA, esiRNA human CTSL1 (esiRNA1)