EMAIL THIS PAGE TO A FRIEND
Molecular nutrition & food research
Dietary squalene supplementation improves DSS-induced acute colitis by downregulating p38 MAPK and NFkB signaling pathways.
PMID 25387687
Abstract
Squalene is a polyunsaturated triterpene, which has exhibited anticancer and antioxidant activities among others. We investigated dietary squalene supplementation effect on an acute colitis model induced by dextran sulfate sodium (DSS) in C57BL/6 mice. Mice were fed from weaning with squalene at 0.02% and 0.1%. After 4 weeks, mice were exposed to 3% DSS for 5 days developing acute colitis. After DSS removal (5 days), colons were histological and biochemically processed. Our results showed that dietary squalene treatment exerts anti-inflammatory action in DSS-induced acute colitis. Western blot revealed that squalene downregulated COX-2 (where COX is cyclooxygenase) and inducible nitric oxide synthase system by inhibition of mitogen-activated protein kinase p38 and the nuclear factor-kappa B signaling pathways, preventing an increase in the cytokines levels. Under our experimental conditions, STAT3 and FOXP3 (where FOXP3 is forkhead box P3) were not modified and the transcriptional regulation of antioxidant and/or detoxifying enzymes, Nrf2 (where Nrf2 is nuclear factor (erythroid-derived 2)-like 2), was reduced in DSS-induced colitis. However, any change could be observed after squalene supplementation. Squalene was able to improve the oxidative events and returned proinflammatory proteins expression to basal levels probably through p38 mitogen-activated protein kinase and nuclear factor-kappa B signaling pathways. However, supplementary studies are needed in order to provide a basis for developing a new dietary supplementation strategy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S3626
Squalene, ≥98%, liquid
C30H50
442785
Squalene, analytical standard
C30H50