EMAIL THIS PAGE TO A FRIEND
Journal of Neuroscience Research
Effect of growth factors and steroid hormones on heme oxygenase and cyclin D1 expression in primary astroglial cell cultures.
PMID 25388553