EMAIL THIS PAGE TO A FRIEND
Mucosal immunology
Intestinal immunopathology is associated with decreased CD73-generated adenosine during lethal infection.
PMID 25389034
Abstract
The ectonucleotidases CD39 and CD73 sequentially degrade the extracellular ATP pool and release immunosuppressive adenosine, thereby regulating inflammatory responses. This control is likely to be critical in the gastrointestinal tract where high levels of ATP are released in particular by commensal bacteria. The aim of this study was therefore to evaluate the involvement of the adenosinergic regulation in the intestine of mice in steady-state conditions and on acute infection with Toxoplasma gondii. We show that both conventional (Tconv) and regulatory (Treg) CD4(+) T lymphocytes express CD39 and CD73 in the intestine of naive mice. CD73 expression was downregulated during acute infection with T. gondii, leading to impaired capacity to produce adenosine. Interestingly, the expression of adenosine receptors was maintained and treatment with receptor agonists limited immunopathology and dysbiosis, suggesting that the activation of adenosine receptors may constitute an efficient approach to control intestinal inflammation associated with decreased ectonucleotidase expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5134
2-Chloroadenosine
C10H12ClN5O4
D9766
Dipyridamole, ≥98% (HPLC)
C24H40N8O4