EMAIL THIS PAGE TO A FRIEND
JAMA dermatology
Binding of the trichodysplasia spinulosa-associated polyomavirus small T antigen to protein phosphatase 2A: elucidation of a potential pathogenic mechanism in a rare skin disease.
PMID 25389794