EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
High-cost generic drugs--implications for patients and policymakers.
PMID 25390739
Abstract
Some older generic drugs have become very expensive, owing to factors including drug shortages, supply disruptions, and consolidations in the generic-drug industry. But generics manufacturers that legally obtain a market monopoly can also unilaterally raise prices.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1012553
Albendazole, United States Pharmacopeia (USP) Reference Standard
C12H15N3O2S