EMAIL THIS PAGE TO A FRIEND
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine
PX-12 induces apoptosis in Calu-6 cells in an oxidative stress-dependent manner.
PMID 25391429