EMAIL THIS PAGE TO A FRIEND
Neurochemical Research
Elevation of BDNF exon I-specific transcripts in the frontal cortex and midbrain of rat during spontaneous morphine withdrawal is accompanied by enhanced pCreb1 occupancy at the corresponding promoter.
PMID 25392083