EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Regulation of neuronal gene expression and survival by basal NMDA receptor activity: a role for histone deacetylase 4.
PMID 25392500
Abstract
Neuronal gene expression is modulated by activity via calcium-permeable receptors such as NMDA receptors (NMDARs). While gene expression changes downstream of evoked NMDAR activity have been well studied, much less is known about gene expression changes that occur under conditions of basal neuronal activity. In mouse dissociated hippocampal neuronal cultures, we found that a broad NMDAR antagonist, AP5, induced robust gene expression changes under basal activity, but subtype-specific antagonists did not. While some of the gene expression changes are also known to be downstream of stimulated NMDAR activity, others appear specific to basal NMDAR activity. The genes altered by AP5 treatment of basal cultures were enriched for pathways related to class IIa histone deacetylases (HDACs), apoptosis, and synapse-related signaling. Specifically, AP5 altered the expression of all three class IIa HDACs that are highly expressed in the brain, HDAC4, HDAC5, and HDAC9, and also induced nuclear accumulation of HDAC4. HDAC4 knockdown abolished a subset of the gene expression changes induced by AP5, and led to neuronal death under long-term tetrodotoxin or AP5 treatment in rat hippocampal organotypic slice cultures. These data suggest that basal, but not evoked, NMDAR activity regulates gene expression in part through HDAC4, and, that HDAC4 has neuroprotective functions under conditions of low NMDAR activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GW21169 Anti-APAF1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
PRS2013
Anti-APAF1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS2015
Anti-APAF1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA031373
Anti-APAF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
PRS2437
Anti-NOXA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2900313
Anti-PMAIP1 (AB2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2501211
Anti-PMAIP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA051063
Anti-PMAIP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500895 Monoclonal Anti-APAF-1 antibody produced in mouse, clone 5E11, affinity isolated antibody
WH0000836M2
Monoclonal Anti-CASP3 antibody produced in mouse, clone 4D3, purified immunoglobulin, buffered aqueous solution
C5737 Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder