EMAIL THIS PAGE TO A FRIEND
Journal of endourology
Which is the preferred modality of renal access for a trainee urologist: ultrasonography or fluoroscopy? Results of a prospective randomized trial.
PMID 25393358
Abstract
In percutaneous nephrolithotomy (PCNL), placement of the access tract into the kidney is an important aspect of the procedure and is responsible for the steep learning curve associated with the operation. The aim of the current prospective randomized trial was to assess the duration of radiation exposure along with the safety and efficacy of PCNL done by a trainee, utilizing either ultrasonography (US) or fluoroscopic guidance to obtain access. Sixty-four patients with ≤3 cm renal calculi undergoing single-tract PCNL by trainee urologists (experience of <25 PCNL's) were randomized into US- and fluoroscopic-guided access groups (32 in each). In Group 1 (US guided), puncture was done using a 3.5/5 MHz US probe with a puncture attachment, whereas the triangulation technique with biplanar C-arm fluoroscopy was utilized in group 2 (fluoroscopic guided). Patient demographics, stone parameters, intraoperative characteristics, fluoroscopy durations, and postoperative outcomes were analyzed. Both groups were comparable as far as patient and stone characteristics were concerned. The fluoroscopy exposure during the puncture phase (T2) and overall (T) was significantly lower in the US group at 9.0±20.8 vs 43.8±34.8, (p<0.0001) and 204.3±84 vs 239.9±77.5, (p=0.04). Six patients in group1 required fluoroscopic adjustment (p=0.03). All other intra- and postoperative parameters were similar in both the groups. Both ultrasound and fluoroscopic guidance for renal access are equally safe and feasible in the hands of a trainee urologist. Total fluoroscopy duration and exposure time during puncture were both significantly less in the ultrasound group. Expertise in fluoroscopic-guided access is essential for a novice to effectively achieve access in all possible situations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
457957
Holmium, chips, 99.9% trace metals basis
Ho
693693
Holmium, foil, thickness 1 mm, 99.9% trace rare earth metals basis
Ho
755974
Holmium, dendritic pieces, purified by distillation, 99.99% trace rare earth metals basis
Ho
GF92551259
Holmium, rod, 50mm, diameter 3.8mm, 99%
Ho
GF12230154
Holmium, rod, 50mm, diameter 6.35mm, cast, 99%
Ho
GF39809863
Holmium, wire reel, 100mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF22997188
Holmium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Ho
GF59427513
Holmium, wire reel, 200mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF74648223
Holmium, wire reel, 200mm, diameter 1.0mm, hard, 99.9%
Ho
GF88732693
Holmium, wire reel, 50mm, diameter 0.5mm, as drawn, 99.9%
Ho
GF13376785
Holmium, rod, 200mm, diameter 3.8mm, 99%
Ho
GF28769541
Holmium, rod, 100mm, diameter 6.35mm, cast, 99%
Ho
GF68907011
Holmium, rod, 100mm, diameter 3.8mm, 99%
Ho
GF31028543
Holmium, powder, 5g, max. particle size 500 micron, 99.9%
Ho
GF78567654
Holmium, powder, 10g, max. particle size 500 micron, 99.9%
Ho
GF29991673
Holmium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Ho
GF26625067
Holmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%
Ho
GF42613006
Holmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99%
Ho
GF60938635
Holmium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Ho
GF34764666
Holmium, foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99%
Ho
GF78609640
Holmium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Ho
GF49265253
Holmium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%
Ho
GF07383977
Holmium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Ho
GF85502640
Holmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Ho
GF19613684
Holmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Ho
GF38519376
Holmium, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99%
Ho