EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical Sciences
Liposomes modified with superhydrophilic polymer linked to a nonphospholipid anchor exhibit reduced complement activation and enhanced circulation.
PMID 25393628