EMAIL THIS PAGE TO A FRIEND
Canadian Journal of Physiology and Pharmacology
Pinosylvin at a high concentration induces AMPK-mediated autophagy for preventing necrosis in bovine aortic endothelial cells.
PMID 25393712
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56297
Pinosylvin, ≥97.0% (HPLC)
C14H12O2