EMAIL THIS PAGE TO A FRIEND
Oncology reports
Emerging role of microRNAs in modulating endothelin-1 expression in gastric cancer.
PMID 25394359
Abstract
Endothelin-1 (ET-1) is a small 21-amino acid peptide that is known to exert diverse biological effects on a wide variety of tissues and cell types through its own receptors. The ET-1-ETRA axis is frequently dysfunctional in numerous types of carcinomas, and contributes to the promotion of cell growth and migration. microRNAs (miRNAs) are small non-coding RNAs that play a critical role in carcinogenesis through mRNA degradation or the translational inhibition of cancer-associated protein-coding genes. However, the role of ET-1 and the relationship between ET-1 and miRNAs in gastric cancer remain unknown. Results of the analysis of the database of The Cancer Genome Atlas (TCGA) revealed that ET-1 is significantly overexpressed in gastric cancer cells when compared with its expression in adjacent normal cells. Exogenous ET-1 significantly enhanced gastric cancer cell proliferation, implying that ET-1 plays an oncogenic role in gastric cancer carcinogenesis. Using a luciferase reporter assay we showed that 18 miRNA candidates had a significant silencing effect on ET-1 expression by up to 20% in HEK293T cells. Among them, 5 miRNAs (miR-1, miR-101, miR-125A, miR-144 and let-7c) were shown to be involved in ET-1 silencing through post-transcriptional modulation in gastric cancer. Our data also revealed that DNA hypermethylation contributes to the silenced miR-1 expression in gastric cancer cells. The ectopic expression of miR-1 significantly inhibited AGS cell proliferation by suppressing ET-1 expression. Overall, our study revealed that ET-1 overexpression may be due to DNA hypermethylation resulting in the silencing of miR-1 expression in gastric cancer cells. In addition, we identified several miRNAs as potential modulators for ET-1 in gastric cancer, which may be used as targets for gastric cancer therapy.