EMAIL THIS PAGE TO A FRIEND
Molecular Medicine Reports
Synthesis, characterization and in vitro and in vivo investigation of C?F?-filled poly(lactic-co-glycolic acid) nanoparticles as an ultrasound contrast agent.
PMID 25394467