EMAIL THIS PAGE TO A FRIEND
Journal of biomolecular screening
Tetracycline-based system for controlled inducible expression of group III metabotropic glutamate receptors.
PMID 25394730
Abstract
A stable and inducible expression of metabotropic glutamate receptor type 4, 7, and 8 was obtained in T-REx 293 cells using the tetracycline system. Tetracycline administration to the cell medium resulted in rapid induction and time-dependent expression of mGlu receptors, which also correlates with its functionality in a cAMP accumulation assay. The pharmacological properties of recombinant mGlu receptors were verified using orthosteric and allosteric ligands. Data suggest that the Tet-on inducible system is suitable for functional mGlu receptors' expression and characterization by means of the cAMP accumulation assay. It makes this system a precise, reproducible, and large-scale screening method, as well as a reasonable tool to study signaling properties of mGlu receptors.