EMAIL THIS PAGE TO A FRIEND
Archives of Toxicology
Assessment of the sensitizing potency of preservatives with chance of skin contact by the loose-fit coculture-based sensitization assay (LCSA).
PMID 25395006