EMAIL THIS PAGE TO A FRIEND
Journal of Mass Spectrometry : Jms
Dereplication of Streptomyces sp. AMC 23 polyether ionophore antibiotics by accurate-mass electrospray tandem mass spectrometry.
PMID 25395127