EMAIL THIS PAGE TO A FRIEND
Journal of Medicinal Food
Red ginseng extract promotes the hair growth in cultured human hair follicles.
PMID 25396716
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64139
Ginsenoside Rg3, analytical standard
C42H72O13
SML0184
Ginsenoside Rg3, ≥98% (HPLC)
C42H72O13