EMAIL THIS PAGE TO A FRIEND
Journal of Pharmacology and Experimental Therapeutics
Polythiol-containing, recombinant mannosylated-albumin is a superior CD68+/CD206+ Kupffer cell-targeted nanoantioxidant for treatment of two acute hepatitis models.
PMID 25398242