EMAIL THIS PAGE TO A FRIEND
Clinical and Experimental Pharmacology & Physiology
Levonorgestrel decreases cilia beat frequency of human fallopian tubes and rat oviducts without changing morphological structure.
PMID 25399777