EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Activatable two-photon fluorescence nanoprobe for bioimaging of glutathione in living cells and tissues.
PMID 25399841
Abstract
Glutathione (GSH) serves vital cellular biological functions, and its abnormal levels are associated with many diseases. To better understand its physiological and pathological functions, efficient methods for monitoring of GSH in living systems are desired. Although quite a few small molecule-based and nanomaterial-based one photon fluorescence probes have been reported for GSH, two-photon (TP) probes, especially nanoprobes with good membrane permeability, are more favorable for bioimaging applications, since TP fluorescence imaging can provide improved spatial localization and increased imaging depth. In this work, we for the first time reported a "turn-on" TP fluorescence nanoprobe for efficient detection of GSH in aqueous solutions and TP excited fluorescence imaging of GSH in living cells and tissues. The nanoprobe consists of two-photon mesoporous silica nanoparticles (TP-MSNs) with a large TP excitation action cross-section (Φδ) value of 103 GM and MnO2 nanosheets, which show intense and broad optical absorption and could act as efficient quenchers for TP fluorescence. In the sensing system, the negatively charged MnO2 nanosheets are adsorbed on the positively charged MSNs through electrostatic interaction, resulting in efficient quenching of their fluorescence, with very low background fluorescence observed. The addition of GSH could reduce MnO2 into Mn(2+), lead to the decomposition of the MnO2 nanosheets, and thereby result in remarkable enhancement of both one photon and TP excited fluorescence of the nanosystem. The nanoprobe shows a highly sensitive response to GSH in aqueous solutions, with a detection limit of 200 nM achieved. It also exhibits a high selectivity toward GSH relative to other biomolecules and electrolytes, with good membrane permeability and excellent biocompatibility. The nanoprobe was successfully applied in monitoring the change of the intracellular GSH in living cells and tissues via TP fluorescence imaging, demonstrating its value of practical application in biological systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
706493
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
C9H23NO3Si
741442
(3-Aminopropyl)triethoxysilane, ≥98.0%
C9H23NO3Si
413364
3-(Triethoxysilyl)propyl isocyanate, 95%
C10H21NO4Si
1102974
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
C19H42BrN
52367
Hexadecyltrimethylammonium bromide, for ion pair chromatography
C19H42BrN
36932
Hexadecyltrimethylammonium bromide, analytical standard
C19H42BrN
52370
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
C19H42BrN
07883
Hexadecyltrimethylammonium bromide, puriss. p.a., matrix substance for MALDI-MS
C19H42BrN
221279
Manganese(II) chloride tetrahydrate, ACS reagent, ≥98%
Cl2Mn · 4H2O
M8054
Manganese(II) chloride tetrahydrate, meets USP testing specifications
Cl2Mn · 4H2O
203734
Manganese(II) chloride tetrahydrate, 99.99% trace metals basis
Cl2Mn · 4H2O
M3634
Manganese(II) chloride tetrahydrate, ReagentPlus®, ≥99%
Cl2Mn · 4H2O
M5005
Manganese(II) chloride tetrahydrate, BioReagent, suitable for insect cell culture
Cl2Mn · 4H2O
63535
Manganese(II) chloride tetrahydrate, BioUltra, for molecular biology, ≥99.0% (KT)
Cl2Mn · 4H2O
31422
Manganese(II) chloride tetrahydrate, puriss. p.a., ACS reagent, ≥99%
Cl2Mn · 4H2O
PHR1317
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8
426318
Tetramethylammonium hydroxide solution, ACS reagent
C4H13NO
334901
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
C4H13NO
331635
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
C4H13NO
328251
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
C4H13NO
89680
Toluene, analytical standard
C7H8
PHR1311
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8