EMAIL THIS PAGE TO A FRIEND
Biological chemistry
miR-103 promotes 3T3-L1 cell adipogenesis through AKT/mTOR signal pathway with its target being MEF2D.
PMID 25400071
Abstract
MicroRNAs are small non-coding RNAs that partially bind to the 3' untranslated (3'UTR) regions of target genes in animals and regulate protein production of the target transcripts. MiR-103 has been confirmed to play a critical role in lipid metabolism, however, the target genes and signaling pathway regulated by miR-103 is still unclear. In our experiment, we observed a positive function of miR-103 on the adipogenic differentiation of 3T3-L1 pre-adipocyte. Furthermore, we proved that this function of miR-103 worked through activating AKT/mTOR signal pathway and impairing target gene MEF2D. By inhibiting and over-expressing the MEF2D gene, we found that MEF2D had a negative role in regulating adipocyte key genes, and this function of MEF2D could be impaired by miR-103. In conclusion, we found that miR-103 can promote 3T3-L1 cells differentiation by targeting MEF2D and activating AKT/mTOR signal pathway. These results will shed a light on further study of microRNAs.