EMAIL THIS PAGE TO A FRIEND
The Journal of clinical investigation
Longistatin in tick saliva blocks advanced glycation end-product receptor activation.
PMID 25401185
Abstract
Ticks are notorious hematophagous ectoparasites and vectors of many deadly pathogens. As an effective vector, ticks must break the strong barrier provided by the skin of their host during feeding, and their saliva contains a complex mixture of bioactive molecules that paralyze host defenses. The receptor for advanced glycation end products (RAGE) mediates immune cell activation at inflammatory sites and is constitutively and highly expressed in skin. Here, we demonstrate that longistatin secreted with saliva of the tick Haemaphysalis longicornis binds RAGE and modulates the host immune response. Similar to other RAGE ligands, longistatin specifically bound the RAGE V domain, and stimulated cultured HUVECs adhered to a longistatin-coated surface; this binding was dramatically inhibited by soluble RAGE or RAGE siRNA. Treatment of HUVECs with longistatin prior to stimulation substantially attenuated cellular oxidative stress and prevented NF-κB translocation, thereby reducing adhesion molecule and cytokine production. Recombinant longistatin inhibited RAGE-mediated migration of mouse peritoneal resident cells (mPRCs) and ameliorated inflammation in mouse footpad edema and pneumonia models. Importantly, tick bite upregulated RAGE ligands in skin, and endogenous longistatin attenuated RAGE-mediated inflammation during tick feeding. Our results suggest that longistatin is a RAGE antagonist that suppresses tick bite-associated inflammation, allowing successful blood-meal acquisition from hosts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV41643
Anti-AGER (AB2) antibody produced in rabbit, IgG fraction of antiserum
R5278 Anti-RAGE (Receptor for Advanced Glycation Endproducts) antibody produced in rabbit, affinity isolated antibody
SAB1401326
Anti-RAGE antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2105049
Anti-RAGE, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB1412548
ANTI-TLR4 antibody produced in mouse, clone 4B10, purified immunoglobulin, buffered aqueous solution
SAB1412602
ANTI-TLR4 antibody produced in mouse, clone 3G12, purified immunoglobulin, buffered aqueous solution
PRS3141
Anti-TLR4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB3500765
Anti-TLR4 antibody produced in rabbit, affinity isolated antibody
RAB0007
Human RAGE / AGER ELISA Kit, for serum, plasma, cell culture supernatant and urine
SAB1404475
Monoclonal Anti-TLR4 antibody produced in mouse, clone 3B6, purified immunoglobulin, buffered aqueous solution
SAB1402375
Monoclonal Anti-TLR4 antibody produced in mouse, clone 1H7, purified immunoglobulin, buffered aqueous solution
RAB0008
Mouse RAGE / AGER ELISA Kit, for serum, plasma and cell culture supernatant
RAB0009
Rat RAGE / AGER ELISA Kit, for serum, plasma and cell culture supernatant
AV41642
Anti-AGER (AB1) antibody produced in rabbit, affinity isolated antibody
SAB2104808
Anti-AGER, (N-terminal) antibody produced in rabbit, affinity isolated antibody