PLoS ONE
Cell fate decisions in malignant hematopoiesis: leukemia phenotype is determined by distinct functional domains of the MN1 oncogene.
PMID 25401736
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D27802
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
C6H12N2
290734
Dabco® 33-LV
C6H12N2
SRP3072 IL-11 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
SRP4135 IL-3 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
I5286 IL-3 from mouse, Carrier free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), cell culture tested
SRP3208 IL-3 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
SRP3096 IL-6 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested
I2406 Interleukin-11 human, >97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, cell culture tested
I4144 Interleukin-3 from mouse, IL-3, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
I2786
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, Animal Component Free
I1395
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
H7416 Interleukin-6 human, IL-6, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
69947
MOPS, BioUltra, for molecular biology, ≥99.5% (T)
C7H15NO4S
PHR1187
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
C20H28O2
1674004
Tretinoin, United States Pharmacopeia (USP) Reference Standard
C20H28O2
T1850000
Tretinoin, European Pharmacopoeia (EP) Reference Standard
C20H28O2