EMAIL THIS PAGE TO A FRIEND
Molecular nutrition & food research
Dietary rapeseed/canola-oil supplementation reduces serum lipids and liver enzymes and alters postprandial inflammatory responses in adipose tissue compared to olive-oil supplementation in obese men.
PMID 25403327
Abstract
Obesity is associated with hyperlipidemia, hepatic steatosis, and low-grade inflammation. Studies have shown that MUFA as well as PUFA have beneficial effects on blood lipids and the inflammatory state. This study investigates the effects of a daily supplementation of either 50 g of rapeseed/canola (RA) or olive (OL) oil over 4 wk on serum lipids, serum liver enzymes, and inflammatory gene expression in subcutaneous (s. c.) adipose tissue in obese men. Consuming RA resulted in increased serum n-3 fatty acids and a reduction in total cholesterol, LDL cholesterol, and serum aspartate aminotransferase compared to OL. In s. c. adipose tissue, gene expression of the pro-inflammatory cytokine IL6 was reduced in RA compared to OL. However, after 4 h after a test meal, containing the appropriate oil, white bread, and 400 mL of liquid diet drink (835 kcal in total), gene expression of IL6, IL1B, and EMR1 (egf-like module containing Mucin-like hormone receptor-like 1) was increased in RA and of monocyte chemoattractant protein-1 (CCL2) in both RA and OL. This demonstrates that consuming RA for 4 wk improves serum lipids, liver enzymes, and basal inflammation in s. c. adipose tissue, but it mediates an acute pro-inflammatory response in adipose tissue upon consuming a meal.